Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139269.2 | NP_644675.2 | 4 | 302 | ||
GenBank mRNA | AB298806.1 | - | 2 | 147 | ||
AK039703.1 | - | 4 | 515 | |||
AK042007.1 | - | 4 | 255 | |||
AK052657.1 | - | 4 | 281 | |||
AK163505.1 | - | 4 | 303 | |||
AX775891.1 | - | 4 | 284 | |||
BC024581.1 | - | 4 | 248 | 5065247 |