Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024560.2 | NP_001019731.1 | 4, 5 | 501 | ||
GenBank mRNA | AK037118.1 | - | 492 | |||
AK047234.1 | - | 4, 5 | 476 | |||
AK143441.1 | - | 3, 4 | 613 | |||
BC048922.1 | - | 4, 5 | 501 | 6311864 |