Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
Trpm3
Gene Name:
transient receptor potential cation channel, subfamily M, member 3
Gene Aliases:
6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2
Chromosome Location:
Chr.19: 22138181 - 22989897 on Build GRCm38
UniGene ID:
Mm.131943
Species:
Mouse
siRNA ID
MSS279020
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Trpm3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001035239.2
NP_001030316.1
5
746
NM_001035240.2
NP_001030317.1
5
746
NM_001035241.2
NP_001030318.1
5
746
NM_001035242.1
NP_001030319.1
5
746
NM_001035243.2
NP_001030320.1
5
746
NM_001035244.1
NP_001030321.1
5
794
NM_001035245.1
NP_001030322.1
5
794
NM_001035246.1
NP_001030323.1
5
794
NM_177341.4
NP_796315.2
5
813
GenBank mRNA
AJ544532.1
-
5
746
AJ544533.1
-
5
746
AJ544534.1
-
5
746
AJ544535.1
-
5
746
AJ544536.1
-
5
746
AK044454.1
-
5
815
AK044533.1
-
5
592
AK046908.1
-
5
627
AK051934.1
-
5
592
AK052064.1
-
5
600
AK053673.1
-
5
631
AK077331.1
-
5
626
AK082141.1
-
5
628
AK084402.1
-
5
599
AK142504.1
-
5
627
AK161909.1
-
5
627
AK163890.1
-
5
621
AK173218.1
-
5
817
BC037449.1
-
5
642
BC141400.1
-
5
789
9056013
BC141412.1
-
5
789
9056025
HM064407.1
-
5
838
HM064408.1
-
5
838
HM064409.1
-
5
838
HM064410.1
-
5
838
HM064411.1
-
5
838
JF706722.1
-
5
809
JX644970.1
-
5
838
JX644971.1
-
5
838
JX644972.1
-
5
838
JX644973.1
-
5
838
JX644974.1
-
5
838
JX644975.1
-
5
838
JX644976.1
-
5
838
JX644977.1
-
5
838
JX644978.1
-
5
838
JX644979.1
-
5
838
JX644980.1
-
5
838
JX644981.1
-
5
838
JX644982.1
-
5
838
JX644983.1
-
5
838
JX644984.1
-
5
838
JX644985.1
-
5
838
JX644986.1
-
5
838
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
cation transmembrane transport
cation channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0