Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
Trpm3
Gene Name:
transient receptor potential cation channel, subfamily M, member 3
Gene Aliases:
6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2
Chromosome Location:
Chr.19: 22138181 - 22989897 on Build GRCm38
UniGene ID:
Mm.131943
Species:
Mouse
siRNA ID
MSS279021
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Trpm3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001035239.2
NP_001030316.1
6
866
NM_001035240.2
NP_001030317.1
6
866
NM_001035241.2
NP_001030318.1
6
866
NM_001035242.1
NP_001030319.1
6
866
NM_001035243.2
NP_001030320.1
6
866
NM_001035244.1
NP_001030321.1
6
914
NM_001035245.1
NP_001030322.1
6
914
NM_001035246.1
NP_001030323.1
6
914
NM_177341.4
NP_796315.2
6
933
GenBank mRNA
AJ544532.1
-
6
866
AJ544533.1
-
6
866
AJ544534.1
-
6
866
AJ544535.1
-
6
866
AJ544536.1
-
6
866
AK044454.1
-
6
935
AK044533.1
-
6
712
AK046908.1
-
6
747
AK051934.1
-
6
712
AK052064.1
-
6
720
AK053673.1
-
6
751
AK077331.1
-
6
746
AK082141.1
-
6
748
AK084402.1
-
6
719
AK142504.1
-
6
747
AK161909.1
-
6
747
AK173218.1
-
6
937
BC037449.1
-
6
762
BC141400.1
-
6
909
9056013
BC141412.1
-
6
909
9056025
HM064407.1
-
6
958
HM064408.1
-
6
958
HM064409.1
-
6
958
HM064410.1
-
6
958
HM064411.1
-
6
958
JF706722.1
-
6
929
JX644970.1
-
6
958
JX644971.1
-
6
958
JX644972.1
-
6
958
JX644973.1
-
6
958
JX644974.1
-
6
958
JX644975.1
-
6
958
JX644976.1
-
6
958
JX644977.1
-
6
958
JX644978.1
-
6
958
JX644979.1
-
6
958
JX644980.1
-
6
958
JX644981.1
-
6
958
JX644982.1
-
6
958
JX644983.1
-
6
958
JX644984.1
-
6
958
JX644985.1
-
6
958
JX644986.1
-
6
958
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
cation transmembrane transport
cation channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0