Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081319.1 | NP_001074788.1 | 19 | 3061 | ||
GenBank mRNA | AK048687.1 | - | 1 | 455 | ||
AK173171.1 | - | 18 | 2643 | |||
BC036137.1 | - | 3 | 857 |