Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198108.2 | NP_932776.1 | 3, 4 | 379 | ||
GenBank mRNA | AK154147.1 | - | 3, 4 | 367 | ||
AK159230.1 | - | 3, 4 | 366 | |||
BC023055.1 | - | 3, 4 | 346 | |||
BC057060.1 | - | 2, 3 | 755 | |||
CV563048.1 | - | 3, 4 | 305 |