Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205132.1 | NP_001192061.1 | 11, 12 | 1638 | ||
NM_001205133.1 | NP_001192062.1 | 11, 12 | 1638 | |||
NM_009535.3 | NP_033561.1 | 11, 12 | 2100 | |||
GenBank mRNA | AK035612.1 | - | 11, 12 | 2087 | ||
AK051466.1 | - | 141 | ||||
AK052478.1 | - | 11, 12 | 1590 | |||
AK053141.1 | - | 1, 2 | 76 | |||
AK167420.1 | - | 11, 12 | 1775 | |||
BC010594.1 | - | 11, 12 | 1474 | 3989783 | ||
X67677.1 | - | 11, 12 | 2027 |