Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253805.1 | NP_001240734.1 | 4 | 909 | ||
NM_001253806.1 | NP_001240735.1 | 4 | 634 | |||
NM_001253807.1 | NP_001240736.1 | 3 | 469 | |||
NM_011740.3 | NP_035870.1 | 5 | 1106 | |||
GenBank mRNA | AK083368.1 | - | 4 | 637 | ||
AK145657.1 | - | 4 | 568 | |||
AK146800.1 | - | 4 | 625 | |||
AK150381.1 | - | 4 | 637 | |||
AK151506.1 | - | 2 | 105 | |||
AK151900.1 | - | 4 | 598 | |||
AK162099.1 | - | 4 | 589 | |||
AK167128.1 | - | 4 | 606 | |||
AK187117.1 | - | 2 | 190 | |||
AK201135.1 | - | 2 | 135 | |||
BC050891.1 | - | 5 | 812 | 6419584 | ||
BC089334.1 | - | 4 | 537 | 30361690 | ||
D78647.1 | - | 4 | 557 | |||
D83037.1 | - | 4 | 591 | |||
D87660.1 | - | 4 | 639 | |||
U79231.1 | - | 3 | 504 |