Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253805.1 | NP_001240734.1 | 5 | 1040 | ||
NM_001253806.1 | NP_001240735.1 | 5 | 765 | |||
NM_001253807.1 | NP_001240736.1 | 4 | 600 | |||
NM_011740.3 | NP_035870.1 | 6 | 1237 | |||
GenBank mRNA | AK083368.1 | - | 5 | 768 | ||
AK145657.1 | - | 5 | 699 | |||
AK146800.1 | - | 5 | 756 | |||
AK150381.1 | - | 5 | 768 | |||
AK151506.1 | - | 3 | 236 | |||
AK151900.1 | - | 5 | 729 | |||
AK162099.1 | - | 5 | 720 | |||
AK167128.1 | - | 5 | 737 | |||
AK187117.1 | - | 3 | 322 | |||
BC050891.1 | - | 6 | 943 | 6419584 | ||
BC089334.1 | - | 5 | 668 | 30361690 | ||
D78647.1 | - | 5 | 688 | |||
D83037.1 | - | 5 | 722 | |||
D87660.1 | - | 5 | 770 | |||
U79231.1 | - | 4 | 635 |