Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081078.1 | NP_001074547.1 | 8 | 3290 | ||
GenBank mRNA | AK158042.1 | - | 2 | 1392 |