Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144875.2 | NP_659124.2 | 2 | 279 | ||
GenBank mRNA | AB232625.1 | - | 1 | 12 | ||
AK079752.1 | - | 1 | 150 | |||
AK137928.1 | - | 1 | 399 | |||
BC016133.1 | - | 2 | 264 | 4485029 | ||
BC029056.1 | - | 1 | 53 | 4947517 | ||
BG087764.2 | - | 2 | 278 |