Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC003331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077237.1 | NP_001070705.1 | 3 | 573 | ||
NM_145511.2 | NP_663486.2 | 3 | 573 | |||
GenBank mRNA | AK152022.1 | - | 1 | 89 | ||
AK152955.1 | - | 1 | 89 | |||
BC003331.1 | - | 2 | 228 | 3256634 | ||
CB235715.1 | - | 3 | 271 | |||
DQ077804.2 | - | 2 | 187 |