Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163307.1 | NP_001156779.1 | 4, 5 | 489 | ||
NM_001163308.1 | NP_001156780.1 | 3, 4 | 351 | |||
NM_009545.2 | NP_033571.1 | 6, 7 | 756 | |||
GenBank mRNA | AF483502.1 | - | 2, 3 | 197 | ||
AF483503.1 | - | 2, 3 | 197 | |||
AK142837.1 | - | 3, 4 | 264 | |||
BC016419.1 | - | 4, 5 | 443 | 4458285 | ||
BI991588.1 | - | 3, 4 | 417 | |||
D90085.1 | - | 6, 7 | 604 |