Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291024.1 | NP_001277953.1 | 4 | 685 | ||
NM_009548.3 | NP_033574.2 | 5 | 754 | |||
GenBank mRNA | AK131651.1 | - | 4 | 629 | ||
BC013139.1 | - | 5 | 648 | 4500921 | ||
BC037118.1 | - | 5 | 702 | 4506141 |