Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291024.1 | NP_001277953.1 | 1, 2 | 208 | ||
NM_009548.3 | NP_033574.2 | 1, 2 | 208 | |||
GenBank mRNA | AB013097.1 | - | 1, 2 | 98 | ||
AK131651.1 | - | 1, 2 | 152 | |||
BC013139.1 | - | 1, 2 | 102 | 4500921 | ||
BC037118.1 | - | 1, 2 | 156 | 4506141 | ||
CJ148450.1 | - | 2, 3 | 289 |