Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081259.1 | NP_001074728.1 | 9 | 1690 | ||
GenBank mRNA | AK036315.1 | - | 8 | 1287 | ||
AK044206.1 | - | 1 | 70 | |||
AK165872.1 | - | 9 | 1690 | |||
BC024751.1 | - | 4 | 668 | |||
BC157992.1 | - | 9 | 1658 | 9007305 |