Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam134a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170755.2 | NP_739561.2 | 7 | 921 | ||
GenBank mRNA | AK034667.1 | - | 7 | 923 | ||
AK152268.1 | - | 7 | 636 | |||
AK158940.1 | - | 7 | 691 | |||
AK160400.1 | - | 7 | 875 | |||
AK165957.1 | - | 7 | 900 | |||
AK172093.1 | - | 7 | 918 | |||
AK189209.1 | - | 1 | 30 | |||
AK210767.1 | - | 1 | 30 | |||
BC026519.1 | - | 3 | 272 | |||
BC038286.1 | - | 3 | 307 | 4221441 | ||
BC070406.1 | - | 7 | 913 | 5707319 |