Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gigyf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110212.2 | NP_001103682.1 | 25, 26 | 3503 | ||
NM_146112.4 | NP_666224.3 | 25, 26 | 3521 | |||
GenBank mRNA | AK122337.1 | - | 16, 17 | 2592 | ||
AY176043.1 | - | 25, 26 | 3467 | |||
BC027137.1 | - | 5, 6 | 866 | |||
BC076610.1 | - | 25, 26 | 3466 | |||
BC082811.1 | - | 6, 7 | 977 | |||
BC089036.1 | - | 25, 26 | 3477 | 30695300 |