Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177344.3 | NP_796318.2 | 1 | 288 | ||
GenBank mRNA | AK050191.1 | - | 1 | 273 | ||
AK051471.1 | - | 1 | 222 | |||
AX775749.1 | - | 1 | 276 | |||
BC022606.1 | - | 1 | 256 | 4165766 |