Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153558.1 | NP_705786.1 | 4 | 400 | ||
GenBank mRNA | AY360148.1 | - | 4 | 436 | ||
BC027556.1 | - | 4 | 400 | 1434737 |