Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033294.3 | NP_001028466.2 | 3, 4 | 786 | ||
GenBank mRNA | AK030888.1 | - | 3, 4 | 733 | ||
AK144739.1 | - | 3, 4 | 499 | |||
AK166327.1 | - | 3, 4 | 456 | |||
BC066017.1 | - | 3, 4 | 423 | 6401415 | ||
BC132351.1 | - | 3, 4 | 476 | 40130628 |