Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252568.2 | NP_001239497.1 | 8 | 876 | ||
NM_001252570.1 | NP_001239499.1 | 8 | 566 | |||
NM_001252571.1 | NP_001239500.1 | 7 | 503 | |||
NM_001281829.1 | NP_001268758.1 | 9 | 592 | |||
NM_172267.4 | NP_758471.1 | 9 | 945 | |||
GenBank mRNA | AA620138.1 | - | 5 | 187 | ||
AK005293.1 | - | 9 | 946 | |||
AK138607.1 | - | 9 | 620 | |||
BC039982.1 | - | 9 | 587 | |||
BC132271.1 | - | 9 | 556 | 40130548 | ||
BC132273.1 | - | 9 | 556 | 40130550 | ||
BC145356.1 | - | 8 | 487 | 9053914 | ||
BC171925.1 | - | 4 | 291 | |||
CX235839.1 | - | 7 | 412 |