Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172268.2 | NP_758472.2 | 12 | 1848 | ||
GenBank mRNA | AK122189.1 | - | 12 | 1817 | ||
AK162214.1 | - | 544 | ||||
BC039282.1 | - | 2 | 205 | 4976266 |