Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146119.2 | NP_666231.1 | 2 | 326 | ||
GenBank mRNA | AK033735.1 | - | 2 | 294 | ||
AK046583.1 | - | 2 | 223 | |||
AK051396.1 | - | 2 | 265 | |||
AK146953.1 | - | 2 | 311 | |||
AK149818.1 | - | 2 | 242 | |||
AK149888.1 | - | 2 | 236 | |||
AK150642.1 | - | 2 | 264 | |||
AK152207.1 | - | 2 | 285 | |||
AK153489.1 | - | 2 | 270 | |||
AK170512.1 | - | 2 | 266 | |||
BC027843.1 | - | 2 | 246 | 5343013 |