Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145535.2 | NP_663510.1 | 8 | 905 | ||
GenBank mRNA | AK136572.1 | - | 8 | 861 | ||
AK144452.1 | - | 8 | 870 | |||
AK160483.1 | - | 8 | 863 | |||
BC005556.1 | - | 8 | 896 | 3964815 |