Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153065.3 | NP_694705.2 | 8 | 860 | ||
GenBank mRNA | AK134274.1 | - | 8 | 861 | ||
AK138442.1 | - | 8 | 884 | |||
AK146469.1 | - | 8 | 887 | |||
AK195653.1 | - | 1 | 89 | |||
AK205436.1 | - | 1 | 89 | |||
BC011321.1 | - | 7 | 737 | 4007058 | ||
BC024730.1 | - | 8 | 860 | |||
BC024981.1 | - | 8 | 860 | |||
BC033398.1 | - | 8 | 860 |