Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isg20l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177663.4 | NP_808331.1 | 2 | 977 | ||
GenBank mRNA | AK053778.1 | - | 2 | 978 | ||
AK076747.1 | - | 2 | 699 | |||
AK155979.1 | - | 2 | 981 | |||
AK178586.1 | - | 1 | 269 | |||
BC098326.1 | - | 2 | 964 | 5720443 |