Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253702.1 | NP_001240631.1 | 3, 4 | 1050 | ||
NM_001253703.1 | NP_001240632.1 | 3, 4 | 1050 | |||
NM_153091.3 | NP_694731.1 | 3, 4 | 1050 | |||
GenBank mRNA | AB084518.1 | - | 3, 4 | 464 | ||
AK033865.1 | - | 3, 4 | 482 | |||
AK144075.1 | - | 3, 4 | 490 | |||
AK148099.1 | - | 3, 4 | 752 | |||
AK172102.1 | - | 3, 4 | 1051 | |||
AK172218.1 | - | 3, 4 | 1051 | |||
AK213571.1 | - | 1, 2 | 55 | |||
AK217761.1 | - | 1, 2 | 55 | |||
AK219361.1 | - | 1, 2 | 55 | |||
BC024473.1 | - | 3, 4 | 441 | |||
BC058781.1 | - | 3, 4 | 475 | 6311899 | ||
BC107394.2 | - | 3, 4 | 475 | 40058131 | ||
BC145950.1 | - | 3, 4 | 542 | 40131253 |