Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146136.1 | NP_666248.1 | 3 | 327 | ||
GenBank mRNA | AK045797.1 | - | 3 | 313 | ||
AK049873.1 | - | 3 | 390 | |||
AK052779.1 | - | 3 | 326 | |||
AK085492.1 | - | 3 | 324 | |||
AK136837.1 | - | 3 | 326 | |||
BC025441.1 | - | 3 | 315 | 5043709 | ||
BC026596.1 | - | 3 | 327 | 4975010 |