Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045807.1 | NP_001039272.1 | 1 | 2183 | ||
GenBank mRNA | AK048987.1 | - | 1418 | |||
AK077048.1 | - | 1 | 2571 | |||
AK166271.1 | - | 1 | 1946 | |||
AK168242.1 | - | 1 | 1944 | |||
AK168936.1 | - | 1 | 2068 | |||
AK220162.1 | - | 1 | 1995 | |||
BC051409.1 | - | 1 | 1937 | |||
BC057038.1 | - | 1 | 1947 | |||
BC080828.1 | - | 1 | 1928 | |||
BC120590.1 | - | 1 | 2183 | 40129513 | ||
BC137741.1 | - | 1 | 2183 | 8860760 |