Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172693.3 | NP_766281.1 | 3 | 693 | ||
GenBank mRNA | AB175680.1 | - | 3 | 673 | ||
AK033638.1 | - | 3 | 694 | |||
AK042133.1 | - | 3 | 726 | |||
AK043977.1 | - | 2 | 553 | |||
AK082014.1 | - | 3 | 739 | |||
AK156751.1 | - | 3 | 723 | |||
BC024475.1 | - | 2 | 525 | |||
BC025639.1 | - | 2 | 415 | |||
BC056425.1 | - | 3 | 704 | 5716909 |