Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175472.3 | NP_780681.2 | 3 | 1201 | ||
GenBank mRNA | AF194972.1 | - | 2 | 966 | ||
AK031299.1 | - | 1 | 594 | |||
AK041821.1 | - | 3 | 1207 | |||
AK049279.1 | - | 1 | 190 | |||
AK220327.1 | - | 1 | 449 | |||
BC145507.1 | - | 2 | 852 | |||
BC150791.1 | - | 2 | 852 | 9087702 |