Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146152.3 | NP_666264.1 | 2 | 1276 | ||
GenBank mRNA | AK052257.1 | - | 2 | 1277 | ||
AK129200.1 | - | 2 | 1259 | |||
AK149978.1 | - | 2 | 1259 | |||
BC031823.1 | - | 2 | 675 | 4188768 |