Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112744.1 | NP_001106215.1 | 12 | 1856 | ||
GenBank mRNA | AK042195.1 | - | 12 | 1852 | ||
AK153708.1 | - | 12 | 2017 | |||
BC020030.1 | - | 9 | 1017 | |||
BC026661.1 | - | 4 | 869 | |||
BC044805.1 | - | 11 | 3270 | |||
BC095966.1 | - | 10 | 1153 | |||
BC137847.1 | - | 12 | 1897 | 8860866 | ||
BC144970.1 | - | 12 | 1897 | 9053518 | ||
BC144971.1 | - | 12 | 1920 |