Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9430015G10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145557.3 | NP_663532.2 | 4 | 277 | ||
NM_177205.1 | NP_796179.1 | 4 | 427 | |||
GenBank mRNA | AK029276.1 | - | 4 | 291 | ||
AK034620.1 | - | 4 | 237 | |||
BC004010.1 | - | 4 | 297 | 3493145 | ||
BQ830972.1 | - | 262 | ||||
BY727027.1 | - | 4 | 406 | |||
CO803636.1 | - | 4 | 393 |