Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198305.2 | NP_938047.1 | 9, 10 | 1538 | ||
GenBank mRNA | AK154523.1 | - | 9, 10 | 1476 | ||
AK170825.1 | - | 9, 10 | 1476 | |||
AY423764.1 | - | 9, 10 | 1516 | |||
BC058738.1 | - | 3, 4 | 307 | |||
BC139309.1 | - | 9, 10 | 1479 | 8862331 |