Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc5g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177677.3 | NP_808345.1 | 4 | 560 | ||
GenBank mRNA | AK076638.1 | - | 4 | 561 | ||
BC089502.1 | - | 4 | 539 | 6774833 |