Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015039.1 | NP_001015039.1 | 12, 13 | 2769 | ||
GenBank mRNA | AK036334.1 | - | 12, 13 | 2773 | ||
AK129414.1 | - | 12, 13 | 2769 | |||
BC042774.1 | - | 5, 6 | 700 | |||
BC139050.1 | - | 12, 13 | 2710 | 8862072 | ||
BC139051.1 | - | 12, 13 | 2710 | 8862073 |