Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173765.3 | NP_776126.1 | 16 | 3298 | ||
GenBank mRNA | AB095954.1 | - | 16 | 3315 | ||
AK136849.1 | - | 16 | 3298 | |||
AK148405.1 | - | 15 | 3124 | |||
BC128330.1 | - | 15 | 3170 | 40110405 |