Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172713.2 | NP_766301.1 | 21, 22 | 2153 | ||
GenBank mRNA | AK029881.1 | - | 21, 22 | 2155 |