Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115010.1 | NP_001108482.1 | 2 | 330 | ||
NM_172714.4 | NP_766302.2 | 2 | 198 | |||
GenBank mRNA | AK028016.1 | - | 2 | 336 | ||
AK030894.1 | - | 2 | 332 | |||
AK134320.1 | - | 2 | 196 | |||
AK220225.1 | - | 2 | 193 |