Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174870.3 | NP_777359.3 | 3 | 1451 | ||
GenBank mRNA | AF349043.1 | - | 2 | 1040 | ||
AK085280.1 | - | 3 | 1160 | |||
AK128888.1 | - | 3 | 1171 | |||
AK143528.1 | - | 3 | 1179 | |||
AY093420.1 | - | 3 | 1371 | |||
BC022130.1 | - | 3 | 1140 | 5098236 | ||
BC025824.1 | - | 3 | 1257 | 5100719 | ||
BC031296.1 | - | 3 | 1164 | |||
BC032151.1 | - | 3 | 1179 | 5055098 |