Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172730.2 | NP_766318.2 | 6 | 630 | ||
GenBank mRNA | AK039818.1 | - | 6 | 609 | ||
AK046253.1 | - | 2 | 2475 | |||
AK081505.1 | - | 5 | 3275 | |||
AK156259.1 | - | 6 | 613 | |||
BC016607.1 | - | 4 | 398 | |||
BC051147.1 | - | 6 | 624 | 6595828 |