Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172731.2 | NP_766319.2 | 8, 9 | 3043 | ||
GenBank mRNA | AK220153.1 | - | 9, 10 | 3517 | ||
BC042732.1 | - | 4, 5 | 460 | |||
BC060664.1 | - | 8, 9 | 2506 | |||
BC145502.1 | - | 9, 10 | 2860 | 9054065 | ||
BC150786.1 | - | 8, 9 | 2992 | 9087697 |