Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145573.2 | NP_663548.2 | 1, 2 | 138 | ||
GenBank mRNA | AK077677.1 | - | 1, 2 | 139 | ||
AK155447.1 | - | 1, 2 | 145 | |||
AK170480.1 | - | 1, 2 | 138 | |||
BC019964.1 | - | 1, 2 | 116 | 4158657 |