Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172738.3 | NP_766326.2 | 5 | 1148 | ||
GenBank mRNA | AK033116.1 | - | 5 | 1124 | ||
BC055688.1 | - | 5 | 1117 | 6402770 |