Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003393.1 | NP_001003393.1 | 1, 2 | 54 | ||
GenBank mRNA | AY672115.1 | - | 1, 2 | 54 | ||
BC147097.1 | - | 1, 2 | 54 | 9056138 |