Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290565.1 | NP_001277494.1 | 4 | 591 | ||
NM_153577.2 | NP_705805.1 | 4 | 591 | |||
GenBank mRNA | AA407649.1 | - | 2 | 62 | ||
BC004761.1 | - | 4 | 540 | 3155758 | ||
BC023803.1 | - | 4 | 578 | 5352953 | ||
BC056649.1 | - | 4 | 530 | 5036575 | ||
BE626968.1 | - | 4 | 516 |