Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2210018M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172280.2 | NP_758484.2 | 19 | 3326 | ||
GenBank mRNA | AK008751.1 | - | 6 | 1309 | ||
AK079153.1 | - | 1 | 496 | |||
BC020109.1 | - | 4 | 827 | |||
BC089304.1 | - | 19 | 3326 | 6854147 |