Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278943.1 | NP_001265872.1 | 11 | 1622 | ||
NM_001278944.1 | NP_001265873.1 | 14 | 2091 | |||
GenBank mRNA | AK144723.1 | - | 11 | 1623 | ||
AK161253.1 | - | 3 | 374 | |||
AY243472.1 | - | 14 | 2090 |